Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cacng1
Entrez Gene ID
496444
Gene Name
calcium channel, voltage-dependent, gamma subunit 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016686059: 2354 - 4728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011035.1 NP_001011035.1 - 510 53
GenBank mRNA BC084053.1 - - 510 53

View Full Product Details