Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gins4
Entrez Gene ID
496455
Gene Name
GINS complex subunit 4 (Sld5 homolog)
Gene Aliases
sld5
Chromosome Location
Chr.3: 119873737 - 119883065 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011045.1 NP_001011045.1 5 - 6 497 76
GenBank mRNA BC084169.1 - 5 - 6 497 76

View Full Product Details