Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hba1
Entrez Gene ID
394454
Gene Name
hemoglobin subunit alpha 1
Gene Aliases
hba-a
Chromosome Location
Chr.9: 36989551 - 36990596 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203529.1 NP_988860.1 1 - 2 137 61
GenBank mRNA BC088005.1 - 1 - 2 119 61
M32454.1 - 1 - 2 137 61

View Full Product Details