Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
emc4
Entrez Gene ID
394471
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
tmem85
Chromosome Location
Chr.1: 130562080 - 130568579 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203545.1 NP_988876.1 4 - 5 429 63
XM_012958022.2 XP_012813476.1 4 - 5 480 63
XM_012958023.2 XP_012813477.1 5 - 6 652 63
GenBank mRNA BC059737.1 - 4 - 5 429 63

View Full Product Details