Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npm1
Entrez Gene ID
394478
Gene Name
nucleophosmin (nucleolar phosphoprotein B23, numatrin)
Gene Aliases
b23, npm, numatrin
Chromosome Location
Chr.3: 9664012 - 9674013 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203552.1 NP_988883.1 8 - 9 735 72
GenBank mRNA BC059736.1 - 8 - 9 735 72
CR761659.1 - 8 - 9 698 72

View Full Product Details