Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
id2
Entrez Gene ID
394480
Gene Name
inhibitor of DNA binding 2
Gene Aliases
gig8, id2a, id2h, xid2
Chromosome Location
Chr.5: 133400308 - 133404616 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203554.1 NP_988885.1 2 - 3 505 74
GenBank mRNA BC059758.1 - 2 - 3 505 74
CR855815.1 - 2 - 3 435 74

View Full Product Details