Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
degs3
Entrez Gene ID
394549
Gene Name
degenerative spermatocyte homolog 3, lipid desaturase
Gene Aliases
-
Chromosome Location
Chr.8: 50240746 - 50255696 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203621.1 NP_988952.1 2 - 3 150 76
GenBank mRNA BC061313.1 - 2 - 3 150 76
CR760077.2 - 1 - 2 154 76

View Full Product Details