Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acat2
Entrez Gene ID
394562
Gene Name
acetyl-CoA acetyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.5: 54222226 - 54235888 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203634.1 NP_988965.1 8 - 9 1108 95
GenBank mRNA BC061429.1 - 8 - 9 1108 95

View Full Product Details