Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pus3
Entrez Gene ID
394566
Gene Name
pseudouridine synthase 3
Gene Aliases
-
Chromosome Location
Chr.9: 8191820 - 8195532 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203638.1 NP_988969.1 4 - 5 1140 84
XM_012970245.1 XP_012825699.1 4 - 5 1167 84
XM_012970246.1 XP_012825700.1 4 - 5 1165 84
XM_012970247.2 XP_012825701.1 4 - 5 1172 84
XM_018097064.1 XP_017952553.1 4 - 5 1109 84
GenBank mRNA BC061416.1 - 4 - 5 1140 84
CR761247.2 - 4 - 5 1183 84
CR848155.2 - 2 - 3 256 84

View Full Product Details