Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
upk3a
Entrez Gene ID
394575
Gene Name
uroplakin 3A
Gene Aliases
up3a, upiii, upiiia, upk3, xUPIII
Chromosome Location
Chr.Un|NW_016683675: 37375 - 42025 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203647.1 NP_988978.2 2 - 3 493 104
GenBank mRNA BC061365.1 - 2 - 3 493 104

View Full Product Details