Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snu13
Entrez Gene ID
394590
Gene Name
small nuclear ribonucleoprotein 13
Gene Aliases
fa-1, fa1, hoip, nhp2l1, nhpx, otk27, snrnp15-5, spag12, ssfa1
Chromosome Location
Chr.4: 103932079 - 103934203 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203663.1 NP_988994.1 2 - 3 221 74
GenBank mRNA BC061279.1 - 2 - 3 221 74
CR760372.2 - 2 - 3 177 74
CR761482.2 - 2 - 3 177 74

View Full Product Details