Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab35
Entrez Gene ID
394615
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
h-ray, rab1c, ray
Chromosome Location
Chr.1: 142916537 - 142931569 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203688.1 NP_989019.1 1 - 2 122 87
GenBank mRNA BC061434.1 - 1 - 2 122 87
CR760193.2 - 1 - 2 81 87

View Full Product Details