Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srprb
Entrez Gene ID
394639
Gene Name
SRP receptor subunit beta
Gene Aliases
apmcf1
Chromosome Location
Chr.5: 126348155 - 126354914 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203711.2 NP_989042.2 2 - 3 308 90
GenBank mRNA BC061370.1 - 2 - 3 276 90
CR848115.2 - 2 - 3 236 90
CX416492.2 - 2 - 3 304 90
CX445223.2 - 2 - 3 308 90

View Full Product Details