Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adss
Entrez Gene ID
394644
Gene Name
adenylosuccinate synthase
Gene Aliases
adeh, adss-a, adss-b, adss2-a, adss2-b
Chromosome Location
Chr.5: 60469229 - 60504980 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203716.1 NP_989047.1 11 - 12 1309 99
XM_012962836.2 XP_012818290.1 11 - 12 1444 99
GenBank mRNA BC061354.1 - 11 - 12 1309 99
CR761467.2 - 11 - 12 1176 99

View Full Product Details