Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkbp6
Entrez Gene ID
394649
Gene Name
FK506 binding protein 6
Gene Aliases
-
Chromosome Location
Chr.2: 37477949 - 37497615 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203721.1 NP_989052.1 4 - 5 525 95
XM_018091140.1 XP_017946629.1 4 - 5 550 95
GenBank mRNA BC061340.1 - 4 - 5 525 95

View Full Product Details