Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tardbp
Entrez Gene ID
394651
Gene Name
TAR DNA binding protein
Gene Aliases
TDP-43, tdp43
Chromosome Location
Chr.7: 39618757 - 39629202 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244758.1 NP_001231687.1 2 - 3 440 105
NM_203723.2 NP_989054.2 2 - 3 440 105
XM_012966272.2 XP_012821726.1 2 - 3 426 105
XM_012966273.2 XP_012821727.1 2 - 3 429 105
GenBank mRNA BC061336.1 - 2 - 3 395 105
CN089905.1 - 2 - 3 442 105
CR762161.2 - 2 - 3 339 105
DN077197.1 - 2 - 3 339 105

View Full Product Details