Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iscu
Entrez Gene ID
394688
Gene Name
iron-sulfur cluster assembly enzyme
Gene Aliases
isu2, nifu, nifun
Chromosome Location
Chr.1: 132848689 - 132856140 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203757.1 NP_989088.1 3 - 4 345 93
GenBank mRNA BC062501.1 - 3 - 4 345 93
CR855562.1 - 3 - 4 321 93

View Full Product Details