Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nt5c1a
Entrez Gene ID
394700
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
-
Chromosome Location
Chr.5: 137993184 - 138003602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203765.1 NP_989096.1 5 - 6 899 70
GenBank mRNA BC062488.1 - 5 - 6 899 70

View Full Product Details