Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcaf7
Entrez Gene ID
394701
Gene Name
DDB1 and CUL4 associated factor 7
Gene Aliases
wdr68
Chromosome Location
Chr.10: 7924157 - 7930843 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203766.1 NP_989097.1 3 - 4 482 83
GenBank mRNA BC062486.1 - 3 - 4 482 83

View Full Product Details