Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ak8
Entrez Gene ID
394708
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, c9orf98, ddx31
Chromosome Location
Chr.8: 7018718 - 7093255 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203773.1 NP_989104.1 8 - 9 765 77
GenBank mRNA BC062516.1 - 8 - 9 765 77

View Full Product Details