Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cops8
Entrez Gene ID
394711
Gene Name
COP9 signalosome subunit 8
Gene Aliases
cop9, cops8-a, cops8-b, cops8.2, csn8, sgn8
Chromosome Location
Chr.9: 48492009 - 48499098 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203775.1 NP_989106.1 5 - 6 474 64
XM_012970259.2 XP_012825713.1 5 - 6 552 64
GenBank mRNA BC062497.1 - 5 - 6 474 64
CR762193.2 - 5 - 6 446 64

View Full Product Details