Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm22
Entrez Gene ID
394713
Gene Name
RNA binding motif protein 22
Gene Aliases
-
Chromosome Location
Chr.6: 83114084 - 83126795 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203777.1 NP_989108.1 2 - 3 129 70
GenBank mRNA BC062518.1 - 2 - 3 129 70
CR761977.1 - 2 - 3 123 70

View Full Product Details