Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdo1
Entrez Gene ID
394737
Gene Name
cysteine dioxygenase type 1
Gene Aliases
-
Chromosome Location
Chr.1: 163334305 - 163350115 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203801.1 NP_989132.1 2 - 3 284 89
GenBank mRNA BC061333.1 - 2 - 3 284 89
CR761233.2 - 2 - 3 283 89

View Full Product Details