Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
etfb
Entrez Gene ID
394759
Gene Name
electron transfer flavoprotein subunit beta
Gene Aliases
-
Chromosome Location
Chr.7: 113344248 - 113373375 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203823.1 NP_989154.1 5 - 6 798 86
GenBank mRNA BC061629.1 - 5 - 6 798 86
BC080971.1 - 5 - 6 798 86
CR942746.2 - 5 - 6 793 86

View Full Product Details