Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bax
Entrez Gene ID
394793
Gene Name
BCL2-associated X protein
Gene Aliases
bax-A, xBax, xlbax
Chromosome Location
Chr.7: 103660655 - 103675501 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203854.1 NP_989185.1 5 - 6 568 79
GenBank mRNA BC063201.1 - 5 - 6 568 79
CR855728.2 - 5 - 6 621 79

View Full Product Details