The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203895.1 | NP_989226.1 | 3 - 4 | 323 | 92 |
XM_012956664.2 | XP_012812118.1 | 3 - 4 | 390 | 92 | |
GenBank mRNA | BC063332.1 | - | 3 - 4 | 323 | 92 |
CR760900.2 | - | 2 - 3 | 303 | 92 | |
CR926239.1 | - | 3 - 4 | 406 | 92 |