Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tktl2
Entrez Gene ID
394835
Gene Name
transketolase-like 2
Gene Aliases
-
Chromosome Location
Chr.8: 14469129 - 14493394 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203896.1 NP_989227.1 7 - 8 993 99
GenBank mRNA BC063226.1 - 7 - 8 993 99

View Full Product Details