Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhrs13
Entrez Gene ID
394935
Gene Name
dehydrogenase/reductase (SDR family) member 13
Gene Aliases
-
Chromosome Location
Chr.2: 26405505 - 26413564 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203980.1 NP_989311.1 3 - 4 390 91
GenBank mRNA BC063926.1 - 3 - 4 390 91
CR926297.2 - 3 - 4 442 91

View Full Product Details