Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cfd
Entrez Gene ID
394945
Gene Name
complement factor D (adipsin)
Gene Aliases
adipsin, ami, df, pfd
Chromosome Location
Chr.1: 81871686 - 81875595 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203989.1 NP_989320.1 3 - 4 373 96
GenBank mRNA BC064208.1 - 3 - 4 373 96

View Full Product Details