Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sult6b1
Entrez Gene ID
394949
Gene Name
sulfotransferase family 6B member 1
Gene Aliases
-
Chromosome Location
Chr.5: 48750369 - 48852749 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203993.1 NP_989324.1 5 - 6 693 119
XM_018093825.1 XP_017949314.1 5 - 6 737 119
XM_018093826.1 XP_017949315.1 5 - 6 711 119
GenBank mRNA BC064180.1 - 5 - 6 693 119
CR761762.2 - 5 - 6 684 119

View Full Product Details