Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hnrnpk
Entrez Gene ID
394952
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
hnrpk
Chromosome Location
Chr.1: 122022548 - 122035494 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203996.1 NP_989327.1 11 - 12 1034 82
XM_012958472.2 XP_012813926.1 11 - 12 1127 82
GenBank mRNA BC064170.1 - 11 - 12 1034 82
CR760855.2 - 11 - 12 978 82

View Full Product Details