Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sdhd
Entrez Gene ID
394986
Gene Name
succinate dehydrogenase complex subunit D, integral membrane protein
Gene Aliases
CybS, cbt1, pgl, pgl1, sdh4, sdhd-a, sdhd-b
Chromosome Location
Chr.7: 74235918 - 74241686 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204026.1 NP_989357.1 1 - 2 91 81
GenBank mRNA BC064178.1 - 1 - 2 91 81
CR848414.2 - 1 - 2 57 81

View Full Product Details