Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftmt
Entrez Gene ID
448718
Gene Name
ferritin mitochondrial
Gene Aliases
-
Chromosome Location
Chr.4: 13612954 - 13618815 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005135.1 NP_001005135.1 2 - 3 331 102
GenBank mRNA BC077674.1 - 2 - 3 331 102
CT027944.2 - 2 - 3 295 102

View Full Product Details