Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snrpe
Entrez Gene ID
448738
Gene Name
small nuclear ribonucleoprotein polypeptide E
Gene Aliases
b-raf, sm-e, sme
Chromosome Location
Chr.4: 67253862 - 67258438 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005147.2 NP_001005147.1 2 - 3 130 84
GenBank mRNA BC077697.1 - 2 - 3 132 84
BC159375.1 - 2 - 3 149 84
CR761996.2 - 2 - 3 86 84

View Full Product Details