Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rcan1
Entrez Gene ID
448016
Gene Name
regulator of calcineurin 1
Gene Aliases
adapt78, csp1, dsc1, dscr1, mcip1, rcn1
Chromosome Location
Chr.2: 10390460 - 10396045 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005434.1 NP_001005434.1 1 - 2 308 109
GenBank mRNA BC074532.1 - 1 - 2 308 109
CR760787.2 - 2 - 3 502 109

View Full Product Details