Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
krtcap3
Entrez Gene ID
448077
Gene Name
keratinocyte associated protein 3
Gene Aliases
kcp3
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005626.1 NP_001005626.1 - 513 70
GenBank mRNA BC074603.1 - - 513 70
CR855774.2 - - 70

View Full Product Details