Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nt5m
Entrez Gene ID
448172
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.9: 9979253 - 9987148 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005673.2 NP_001005673.2 2 - 3 663 70
XM_012970294.2 XP_012825748.1 3 - 4 824 70
GenBank mRNA BC074711.1 - 2 - 3 663 70
EG653972.1 - 2 - 3 286 70

View Full Product Details