Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nudt4
Entrez Gene ID
448180
Gene Name
nudix hydrolase 4
Gene Aliases
dipp2, dipp2alpha, dipp2beta
Chromosome Location
Chr.3: 46614495 - 46642902 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005679.1 NP_001005679.1 1 - 2 344 63
GenBank mRNA BC075099.1 - 1 - 2 344 63

View Full Product Details