Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctss
Entrez Gene ID
448203
Gene Name
cathepsin S
Gene Aliases
ctss-a, ctss-b, ctss.1
Chromosome Location
Chr.8: 106367714 - 106385469 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005695.1 NP_001005695.1 6 - 7 878 70
GenBank mRNA BC075261.1 - 6 - 7 878 70

View Full Product Details