Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nipsnap3a
Entrez Gene ID
448244
Gene Name
nipsnap homolog 3A
Gene Aliases
nipsnap3a-a, nipsnap3a-b, nipsnap4, tassc
Chromosome Location
Chr.1: 107770807 - 107786255 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005718.1 NP_001005718.1 3 - 4 574 69
XM_012959387.2 XP_012814841.1 3 - 4 710 69
GenBank mRNA BC075307.1 - 3 - 4 574 69

View Full Product Details