Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stxbp6
Entrez Gene ID
448250
Gene Name
syntaxin binding protein 6
Gene Aliases
amisyn, hspc156
Chromosome Location
Chr.8: 91620509 - 91656185 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005719.1 NP_001005719.1 4 - 5 822 60
GenBank mRNA BC075314.1 - 4 - 5 822 60

View Full Product Details