Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcolce2
Entrez Gene ID
448281
Gene Name
procollagen C-endopeptidase enhancer 2
Gene Aliases
-
Chromosome Location
Chr.5: 116287320 - 116316491 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005804.1 NP_001005804.1 5 - 6 847 99
GenBank mRNA BC075353.1 - 5 - 6 847 99
CR760811.2 - 5 - 6 801 99

View Full Product Details