Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nif
Entrez Gene ID
448000
Gene Name
low molecular weight neuronal intermediate filament
Gene Aliases
CTDSPL, XNIF, ina, inexa
Chromosome Location
Chr.3: 112174262 - 112180680 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006106.1 NP_001006106.1 3 - 4 1227 89
GenBank mRNA BC074515.1 - 3 - 4 1227 89

View Full Product Details