Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab30
Entrez Gene ID
448082
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
rsb30
Chromosome Location
Chr.2: 168399065 - 168442794 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006108.1 NP_001006108.1 3 - 4 342 110
XM_012956713.2 XP_012812167.1 2 - 3 244 110
XM_012956714.2 XP_012812168.1 3 - 4 388 110
GenBank mRNA BC074609.1 - 3 - 4 342 110
CT025302.2 - 3 - 4 282 110

View Full Product Details