Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2q2
Entrez Gene ID
448322
Gene Name
ubiquitin conjugating enzyme E2Q family member 2
Gene Aliases
ube2q
Chromosome Location
Chr.3: 80498243 - 80516781 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006695.1 NP_001006696.1 11 - 12 1157 98
GenBank mRNA BC075400.1 - 11 - 12 1157 98
CR761026.2 - 11 - 12 1131 98

View Full Product Details