Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccng1
Entrez Gene ID
448326
Gene Name
cyclin G1
Gene Aliases
ccng
Chromosome Location
Chr.3: 15923374 - 15932510 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006697.1 NP_001006698.1 5 - 6 884 72
GenBank mRNA BC075404.1 - 5 - 6 884 72
CR761487.2 - 5 - 6 893 72

View Full Product Details