The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006714.2 | NP_001006715.1 | 3 - 4 | 259 | 83 |
XM_012966387.2 | XP_012821841.1 | 3 - 4 | 199 | 83 | |
GenBank mRNA | BC075442.1 | - | 3 - 4 | 261 | 83 |