Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab34
Entrez Gene ID
448455
Gene Name
RAB34, member RAS oncogene family
Gene Aliases
rab39, rah
Chromosome Location
Chr.2: 26760596 - 26782313 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006768.1 NP_001006769.1 8 - 9 726 90
GenBank mRNA BC075563.1 - 8 - 9 726 90

View Full Product Details