Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnal4
Entrez Gene ID
448476
Gene Name
dynein axonemal light chain 4
Gene Aliases
pig27
Chromosome Location
Chr.4: 110192815 - 110202490 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006783.1 NP_001006784.1 3 - 4 464 88
GenBank mRNA BC075591.1 - 3 - 4 464 88
CR762240.2 - 2 - 3 218 88

View Full Product Details