Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gch1
Entrez Gene ID
448485
Gene Name
GTP cyclohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.8: 74500215 - 74519103 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006788.1 NP_001006789.1 4 - 5 632 97
GenBank mRNA BC075602.1 - 4 - 5 632 97

View Full Product Details